Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Marking Pens
SCI_ED

Catalog No. S32181 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S32181 Red
S32179 Black
S32180 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S32181

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quick-drying ink has no unpleasant odor.

  • Fast drying—highly resistant to abrasion and water
  • Designed to write on cold and damp surfaces as easily as they write on dry surfaces
  • Will not smear when subjected to water or alcohol during normal use
  • Ink is permanent on plastic, wood, and ceramic
  • Washes off glass, metal, or porcelain surfaces

Order Info

Sold in packs of 10

Specifications

Product Type Lab Permanent Marker
Tip Style Fine
Color Red
Sterility Non-sterile
Latex Free Yes
For Use With (Application) Used to make permanent markings on plastic, metal, or porcelain labware
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.