Learn More
Fisherbrand™ Medium-Pointed Dissecting Forceps
Fisherbrand Medium-Pointed Dissection Forceps are used for grasping tissue and materials during dissection. ASTM™ standard stainless-steel
 View more versions of this product
Catalog No. 08-885
Add to cart
Description
Description
Fisherbrand Medium-Pointed Dissection Forceps are used for grasping tissue and materials during dissection. ASTM™ standard stainless-steel
  • Fisherbrand™ Medium-Pointed Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Tips grasp tissues and materials
  • Tips meet precisely when closed
  • Patterned-grip handles with guide pins (08-882 and 08-885)
  • Smooth handle without guide pins (08-900)
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Specifications
Yes
Sterling Silver
Grasping of tissues and materials
4.5 in.
Straight, serrated
Medium Pointed Forcep
ASTM
High Polish, Mirror Finish
Yes
11.4cm
Nickel-plated Steel