Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Microcentrifuge Tubes Accessory Tube Rack

Catalog No. 0566468
Change view
Click to view available options
Positions:
40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
05-664-68 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-664-68 Supplier Fisherbrand™ Supplier No. 4214
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

40-Place, aluminum tube rack

Specifications

Product Type Microcentrifuge Tube Rack
Autoclavable Autoclavable
Color Silver
Material Aluminum
Positions 40
For Use With (Equipment) Conical microcentrifuge tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.