Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Square Microscope Slide Labels
GREENER_CHOICESDP

Catalog No. 15908 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
0.875 x 0.875 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
15908 0.875 x 0.875 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended to be used to label microscope slides. Designed to be individually fed into a laser or ink jet printer.

  • Individual labels measure 7/8 x 7/8 in.
  • 40 labels to a sheet, sold in packs of 1,000 labels (25 sheets per pack)

Specifications

Color White
Format Label
For Use With (Application) Use on microscope slides
For Use With (Equipment) Laser and inkjet printer
Quantity 1000/Pk.
Dimensions (L x W) 0.875 x 0.875 in.
Width (English) 0.87 in.
Height (English) 0.87 in.
Height (Metric) 2.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.