Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Metal Multiple Tape Dispenser for 1 in. Tape
GREENER_CHOICESDPSCI_ED

Catalog No. S68154 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Multiple-Roll Tape Dispenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S68154 Multiple-Roll Tape Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68154
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds multiple rolls.

  • Dispenses multiple rolls of tape to have a variety of colors at hand
  • Each roll can be up to 500 in. (12.7m) long

Specifications

Product Type Multiple-Roll Tape Dispenser
Capacity Multiple rolls, each roll can be up to 500 in. L
For Use With (Application) Dispenses multiple rolls of tape to have a variety of colors at hand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.