Learn More
Fisherbrand™ Neutral-Sol™ Chemical Burn First Aid Station
Catalog No. 1003010 Shop All Fisherbrand Products
Catalog No. 10-030-10
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
One 32 oz. (946mL) bottle of Neutral-Sol and wall mounting screws
Instant acid and caustic neutralization
Provides emergency treatment for acid and alkaline chemical burns. Neutralizes chemicals accidentally splashed on skin, eyes, or clothing in seconds. Also provides flushing action for dust.
  • For labs, chemical storage areas, plants, and any areas where chemicals are used
  • Easy-grab, wide-mouth bottle contains Neutral-Sol solution, a sterile 6.9pH phosphate buffer similar to that present in human tissue
  • Bottle securely housed in brilliant green ABS plastic station for fast recognition
Compliance
Complies with OSHA regulations for quick drenching and flushing action.