Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Permaloc™ Formalin Containers
SDP

Catalog No. 22126347 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
120 mL, pre-filled 60 mL
20 mL, pre-filled 10 mL
40 mL, pre-filled 20 mL
60 mL, pre-filled 30 mL
90 mL, pre-filled 45 mL
No. per Case:
128/Cs.
192
324/Cs.
48/Cs.
480/Cs.
768/Cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. per Case
22-126-347 20 mL, pre-filled 10 mL 768/Cs.
22-126-346 20 mL, pre-filled 10 mL 128/Cs.
22-126-350 40 mL, pre-filled 20 mL 48/Cs.
22-126-351 40 mL, pre-filled 20 mL 480/Cs.
22-126-354 90 mL, pre-filled 45 mL 48/Cs.
22-126-352 60 mL, pre-filled 30 mL 324/Cs.
22-126-357 120 mL, pre-filled 60 mL 192
22-126-356 120 mL, pre-filled 60 mL 48/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-126-347

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Double-click locking ensures specimen integrity while guarding against lost specimens through accidental spills

  • Patented double-clicking lock design
  • 1st audible“click”ensures closure
  • 2nd audible“click”achieves full 95kPa certification
  • Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
  • Leak tested and independently 95kPa certified
  • Avoid employee/patient exposure to hazardous chemicals
  • Improve efficiency of specimen transport to the laboratory
  • Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs

Specifications

Product Type Formalin Container
Capacity (Metric) 20 mL, pre-filled 10 mL
Closure Type Double clicking lock design
No. per Case 768/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.