Learn More
Fisherbrand™ PhoneWipe
SDP
Catalog No. 18393 Shop All Fisherbrand Products
Click to view available options
For Use With (Application):
For cleaning and disinfecting telephones, microphones and other shared communications equipment, PhoneWipe is an easy to use, pre-moistened germicidal wiper that kills most germs on hard, nonporous surfaces
Catalog No. For Use With (Application)
18-393 For cleaning and disinfecting telephones, microphones and other shared communications equipment, PhoneWipe is an easy to use, pre-moistened germicidal wiper that kills most germs on hard, nonporous surfaces
1 options
This item is not returnable. View return policy
Catalog No. 18-393
This item is not returnable. View return policy
PhoneWipe is used for cleaning and disinfecting telephones, microphones and other shared communications equipment. The pre-saturated 5 x 7 in. wipes with 40% ethyl alcohol, .5% benzethonium chloride and water kills all nonspore-forming bacterial strains including Staphylococcus Aureus, Pseudomonas Aeruginosa and Salmonella Choleraesuis on hard, nonporous surfaces.
  • Kill all non spore-forming bacterial strains: Staphylococcus aureus, Pseudomonas aeruginosa, Salmonella and choleraesuis on hard non porous surfaces
  • Pre-moistened germicidal wipes
Specifications
Product Type Phone Wipes
For Use With (Application) For cleaning and disinfecting telephones, microphones and other shared communications equipment, PhoneWipe is an easy to use, pre-moistened germicidal wiper that kills most germs on hard, nonporous surfaces