Description
Durable, zero particulate shoe covers repel liquids.
- Constructed of blue polylatex fabric
- With seamed or seamless bottom
- Elastic ankle for comfortable fit
Order Info
Packed in double polybag.
Specifications
Specifications
| Size | Regular |
| Material | Polylatex |
| Latex Free | No |
| Color | Blue |
| Closure Type | Elastic Ankle |
| Sole Type | Seamed Bottom |
| Resistance | Fluid |
| Product Type | Shoe Covers |