Description
- Withstand autoclaving temperatures of up to 175°C
- All-plastic construction eliminates liquid meniscus for easier and more accurate readings
- Octagonal bases improve stability
Specifications
Specifications
| Autoclavable | Autoclavable |
| Max. Temperature | 175°C |
| Scale Color | Clear |
| Base Type | Octagonal |
| Type | Graduated Cylinder |