Learn More
Fisherbrand™ Polypropylene Slide Mailers
Catalog No. 22363901 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100/Pk.
25/Pk.
Closure Type:
End Opening
Side Opening
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Closure Type
22-363-901 25/Pk. Side Opening
22-363-900 100/Pk. End Opening
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-363-901
This item is not returnable. View return policy
Available with side and end opening configurations
  • Sturdy polypropylene
  • Each mailer holds five 3 × 1 in. slides
Specifications
Closure Type Side Opening
Device Holds 3 x 1 in. Slides
Material Polypropylene
Quantity 25/Pk.