Learn More
Fisherbrand™ Polypropylene Tubing 4-Way Connectors
Very durable, autoclavable, and highly resistant to many chemicals.
Description
- Excellent chemical resistance
- 4-Way Tubing Connectors have serrated connecting surfaces to prevent tubing slipping
- Tubing connectors are molded of polypropypene
Specifications
Tubing Connector | |
0.156 in. | |
0.99 in. | |
4mm | |
1.45 in. | |
25 mm | |
Transparent |
Autoclavable | |
Polypropylene | |
25 mm | |
36.8 mm | |
4-way | |
0.99 in. | |
20 psi |