Learn More
Fisherbrand™ Polystyrene Sample Vials
Catalog No. 0334113 Shop All Fisherbrand Products
Catalog No. 03-341-13
High-impact, fast-flow polystyrene single-use sample vials for automated blood cell counters
  • 20mL capacity
  • Optically clear, parallel sides
  • Static-free and deionized to ensure accurate cell and particle counts
  • With leak-resistant snap cap
  • In shrink-wrapped, vacuum-formed trays of 72