Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Insulated Leather Drivers Gloves: Thermal Lined

Catalog No. p-4893617 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-081356 Medium
19-081357 Large
19-081358 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-081356
This item is not returnable. View return policy

Made from top-grain cowhide

Ideal for use in cold weather applications including farming, construction, machine operation, and refrigeration.

  • Straight thumb and shirred elastic back
  • White thermal lining
  • Gunn-cut style
  • Easy size identification by color and cuff

Specifications

Color Natural
Grade Premium
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Medium
Style Gunn
For Use With (Application) Farming, Construction, Machine Operation, and Refrigeration
Glove Cuff Style Open
Hem Color Green
Lining Thermal Lining
Thumb Style Straight
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.