Learn More
Fisherbrand™ Racer Series Crew Chief Eyewear
Catalog No. 190421017 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Silver
Lens Tint:
Clear
Dark Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-042-1017 Black Dark Gray
19-042-1018 Silver Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042-1017
This item is not returnable. View return policy
Exceptional protection and style at an affordable price
  • True dual-lens styling makes this a fashionable choice
  • A soft nose bridge and rubber-tipped side arms offer a comfortable grip Choose from black nylon frames
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1, ANSI Z80.3
Lens Tint Dark Gray
Frame Color Black
Lens Material Polycarbonate