Learn More
Fisherbrand™ Racer Series Qualifier Eyewear
Catalog No. 190421000 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Red
Lens Tint:
Clear
Dark Gray
Flash Mirror
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-042-1000 Black Clear
19-042-1001 Black Dark Gray
19-042-1002 Black Flash Mirror
19-042-1003 Red Clear
19-042-1004 Red Dark Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-042-1000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Offers exceptional protection and style at an affordable price
  • Flared single-lens design provides sleek protection
  • Soft nose bridge and bayonet-style side arms grip lightly but firmly for a comfortable, confident fit
  • Black nylon frames
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Lens Coating Scratch-resistant
Shape Oval
Certifications/Compliance ANSI Z87.1, ANSI Z80.3
Lens Tint Clear
Product Type Safety Glasses
Frame Color Black
Material Nylon (Frame)
Lens Material Polycarbonate