Learn More
Fisherbrand™ Racer Series Qualifier Sport Eyewear
Catalog No. 19181540 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black, Blue
Blue
Silver, Black
Lens Tint:
Blue
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-181-540 Blue Blue
19-181-542 Silver, Black Gray
19-181-541 Black, Blue Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-181-540
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Frameless single lens design with soft nose bridge and temples to grip the face lightly but firmly
  • Frameless single lens design
  • Soft nose bridge and temples to grip the face lightly but firmly
  • Blue lens commonly used for indoor applications with an excessive amount of yellow or sodium vapor light
  • Gray lens minimizes outdoor sunlight and glare Clear lens is excellent for most indoor applications
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Certifications/Compliance ANSI Z87.1, ANSI Z80.3
Lens Tint Blue
Frame Color Blue