Learn More
Fisherbrand™ Racer Series Qualifier Sport Eyewear
Catalog No. 19181542 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black, Blue
Blue
Silver, Black
Lens Tint:
Blue
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-181-542 Silver, Black Gray
19-181-540 Blue Blue
19-181-541 Black, Blue Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-181-542
This item is not returnable. View return policy
Frameless single lens design with soft nose bridge and temples to grip the face lightly but firmly
  • Frameless single lens design
  • Soft nose bridge and temples to grip the face lightly but firmly
  • Blue lens commonly used for indoor applications with an excessive amount of yellow or sodium vapor light
  • Gray lens minimizes outdoor sunlight and glare Clear lens is excellent for most indoor applications
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
Certifications/Compliance ANSI Z87.1, ANSI Z80.3
Lens Tint Gray
Frame Color Silver, Black