Learn More
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
 View more versions of this product
Catalog No. FB5011000
Add to cart
Description
Description
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
  • Capacity scale measures approximate volume
  • Matte-finish ceramic marking spot takes writing and erases easily
Specifications
Specifications
Rubber
Top style: Heavy duty tooled with capacity scale
122mm
Borosilicate Glass
250 to 1000mL
8.5 in.
Wide
Matte finish Ceramic Enamel marking
11
4.8 in.
1000 mL
No
50mL
21.6cm
1.0