Learn More
Epredia™ Ring Slide for CytoFuge™ 2
Catalog No. 22230901 Shop All Epredia Products
Change view
Click to view available options
Type:
Ring Slide
Catalog No. Type
22-230-901 Ring Slide
1 options
This item is not returnable. View return policy
Catalog No. 22-230-901 Supplier Epredia™ Supplier No. ES3025W
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to collect a thin layer of cells in the ringed area
  • Fully compatible with the Statspin™ CytoFuge 2 cytocentrifuge system
  • 7mm diameter single ring
Specifications
Type Ring Slide