Learn More
Fisherbrand™ SAFE-D-Spense™ 24-Hour Urine Container
Catalog No. 0254032 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
3000 mL
Catalog No. Volume (Metric)
02-540-32 3000 mL
1 options
Catalog No. 02-540-32
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
With dripless pour spout
  • Improved engravings and new cap assure precise measuring and dispensing of urine samples
  • Precision-made, fine pour spout allows for transferring small quantities of sample into small containers
  • Captive spout cap prevents cross-contamination and cap loss
  • Amber color for UV protection
  • 3000mL capacity
Specifications
Product Type 24-Hour Urine Specimen Collection Container
Closure Type Pour-spout Cap
Graduated Yes
Material Polyethylene
Color Amber
No. per Case 40/Cs.
Volume (Metric) 3000 mL