Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety Lancets
SDP

Catalog No. 02675165 Shop All Fisherbrand Products
Change view
Click to view available options
High Flow; Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
02-675-165 High Flow; Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-675-165

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Built-in needle control depth (1.8mm) for consistent and accurate sampling results

  • Sterile lancet needle is hidden inside the cartridge and automatically retracts after use
  • Helps protect patient and health professional against accidental needlestick injuries and cross-contamination
  • Proprietary spring technology inhibits access to lancet after use — lancet cannot be reloaded or reused

Specifications

Sterility Sterile
Includes Proprietary spring technology
Flow Type High
Product Type Safety Lancet
For Use With (Application) Helps protect patient and health professional against accidental needlestick injuries and cross-contamination
Depth (Metric) Needle 1.8mm
Depth (English) Needle 0.07 in.
Quantity 150 Pack
Color Yellow
Style Retract
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.