Learn More
Hellma Secondary Spectrophotometric Calibration Standards
Catalog No. 14385945 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Standard Calibration Set
Catalog No. Type
14-385-945 Standard Calibration Set
1 options
Catalog No. 14-385-945
Add to Cart
Add to Cart
Request a Quote
Calibration of the secondary standards is accomplished through the use of a high-performance UV/VIS/NIR spectrophotometer
These standards allow calibration of wavelength and absorbance for almost any spectrophotometer. Especially useful for QC, environmental or pharmaceutical laboratories that require verification for compliance with U.S. EPA or FDA regulations.
  • Instrument checked at fixed intervals with the aid of a set of primary standards from the National Institute of Standards and Technology (NIST™) and is used exclusively for this purpose
  • Because the successful use of the secondary standards is dependent on their proper handling with respect to the spectrophotometer, each set comes complete with extensive handling instructions
  • Standard set is supplied in a storage case with five glass filters
Specifications
Certifications/Compliance US EPA, FDA regulations
Type Standard Calibration Set
For Use With (Equipment) Secondary spectrophotometric calibration
Includes 5 Glass filters, Holmium Oxide Glass filter, NG 11, NG 5, NG 4 and Empty filter mount