Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Securline™ Blood Bands

Catalog No. 1490521 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Red
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-905-21 Yellow
14-905-19 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-905-21

missing translation for 'validMainframeMsg'

Ideal for use in trauma centers

  • An efficient device for crossmatching blood type for transfusion
  • Large labels feature easy-to-read, white information area, pre-printed serial numbers and “carbonless copy“ underneath
  • Labels are removed from the band and adhered to draw tubes, units of blood, other specimen tubes or paperwork
  • Match the serial numbered label on the unit of blood with the patient's blood band before transfusion to prevent fatal errors

Specifications

Includes Large labels
Color Yellow
Product Type Securline™ Blood Band
For Use With (Application) Trauma centers, efficient device for crossmatching Blood type for transfusion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.