Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Securline™ Marker II Pens

Catalog No. 1490535 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-905-35 Red
14-905-30 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-905-35

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specially designed for marking Superfrost™ and frosted microslides and plastic embedding cassettes

Performs well on ceramic, vinyl, paper, porcelain, metal, Mylar™ and other types of glass and plastic. Please test on material before using.

 

  • Ink is resistant to many normal laboratory solutions when used in procedures that do not require abrasive contact
  • Resists a variety of such solvents as alcohol, acetone and xylene
  • Perfect for histology and cytology work
  • Ink will withstand steam or gas sterilization and temperatures from cryogenic to approximately 150°C (238°F)
  • Line width is approximately 0.015 in.
  • Ink dries to the touch almost immediately, but should be allowed to fix for five minutes before processing in solvent baths
  • Black or red

 

Specifications

Product Type Marker II Pen
Color Red
For Use With (Application) Used for marking Superfrost™ and frosted microslides and Plastic embedding cassettes
Quantity 10 Pk.
Width (Metric) Line 0.38 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.