Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Selectfrost™ Microscope Slides

Catalog No. p-3574511 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Burnt Orange
Green
Lilac
Pink
Tan
White
Yellow
Corner Style:
Clipped
Plain
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
12-550-05 Aqua Plain
12-550-01 Blue Plain
12-550-06 Burnt Orange Plain
12-550-02 Green Plain
12-550-07 Lilac Plain
12-550-03 Pink Plain
12-550-08 Tan Plain
12-550-04 Yellow Plain
12-550-003 White Plain
22-037-205 White Clipped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 12-550-05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color coded for easy identification

The Selectfrost microscope slides include colored tabs that allow grouping according to urgency, test, hospital or physician.

  • Autoclavable
  • Available in an array of tab colors
  • Durable, opaque tab is inert to common laboratory chemicals and reagents
  • Does NOT include SPECIMEN orientation key printed below tab
  • Packaged in paperboard box
  • Size: 25 × 75 × 1mm

Specifications

Length (Metric) 25 mm
Frosted Selectfrost Color-coded
Width (Metric) 75 mm
Thickness (Metric) 1 mm
Type Microscope Slide
Color Aqua
Corner Style Plain
Quantity 144/Pk.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.