Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sharps-A-Gator™ Sharps Container for Phlebotomy

Catalog No. 22037970 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
1 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-037-970 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-037-970

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sharps containers for efficient disposal of blood, hypodermic and IV needles

For safe, efficient disposal of blood, hypodermic and IV needles.

  • Bright red, one-piece polypropylene container clearly marked with biohazard label
  • Translucent top has convenient holder for blood tubes with a needle remover connected to the holder
  • Sliding lid can snap into open position, slide fully shut without locking, or permanently lock shut
  • Safe to autoclave or incinerate

Specifications

Capacity (Metric) 0.9 L
Closure Type Lockable
Closure Color Translucent
Capacity (English) 1 qt.
Color Red
Dimensions (L x W x H) 4.25 x 4.25 x 6.25 in.
Disposable Single-use
Height (Metric) 15.9 cm
Length (Metric) 10.8 cm
Quantity 18/Cs.
Width (Metric) 10.8 cm
Autoclavable Autoclavable
For Use With (Application) Facilitates the disconnection of multi-sample needles from needle holders
Width (English) 4.25 in.
Length (English) 4.25 in.
Height (English) 6.25 in.
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.