Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SHUR/Clean™ Active Sloped-Front Fume Hoods for Coverslipping
SDP

Catalog No. 22047203 Shop All Fisherbrand Products
Change view
Click to view available options
Voltage:
100/115 V
220/240 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
22-047-203 100/115 V
22-047-204 220/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-047-203
Add to Cart
Add to Cart
Request a Quote

Bench-top fume hoods to help manage routine manual histological processes

The Sloped-Front Fume Hood features two 174 cfm high-speed fans that pull contaminated air through two 8 x 8 in. prefilters to remove dust and particulate matter and two 8 x 8 in. charcoal filters to provide the protection needed for hazardous fume absorption. Double-hinged front sash provides convenient access for multiple applications.

  • Microprocessor controlled
  • Double-hinged sash
  • Two prefilters for particulate removal and two carbon filters for fume containment
  • Two constant-speed 174 cfm blowers
  • Face velocity: 90 fpm with sash completely down
  • 22 in. fluorescent 13W T-5 bulb
  • External dimensions: 36W x 20D x 28 in.H (91 x 51 x 71cm)
  • Internal dimensions: 34W x 19D x 19 in.H (86 x 48 x 48cm)
  • Weight: 67 lb. (31kg)
  • Model SC-36S-X (for Coverslipping): contains two carbon filters (SC-36S-XF) for xylene fume containment

Specifications

Product Type Ductless Fume Hood
Depth (English) 20 in.
Depth (English) Interior 19 in.
Depth (Metric) 51 cm
Depth (Metric) Interior 48 cm
Dimensions (D x W x H) 19 x 34 x 19 in. (48 x 86 x 48 cm)
For Use With (Application) Coverslipping
Height (English) 28 in.
Height (English) Interior 19 in.
Height (Metric) 71 cm
Height (Metric) Interior 48 cm
Voltage 100/115 V
Weight (English) 67 lb.
Weight (Metric) 31 kg
Width (English) 36 in.
Width (English) Interior 34 in.
Width (Metric) 91 cm
Width (Metric) Interior 86 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.