Learn More
Fisher Healthcare™ SHUR/Clean™ Downdraft Ductless Fume Hood
SDP
Catalog No. 22044653 Shop All Fisherbrand Products
Change view
Click to view available options
Filter Type:
Xylene Absorbing
Catalog No. Filter Type
22-044-653 Xylene Absorbing
1 options
Catalog No. 22-044-653
Add to Cart
Add to Cart
For coverslipping applications utilizing xylene-based mounting mediums
Ductless, compact and portable.
  • Microprocessor control of on/off times and fan speed provides unparalleled efficiencies and robust fume management
  • Fumes are pulled down by a 225cfm fan through a perforated work surface and absorbed by a replaceable activated charcoal cartridge
Order Info
Note: 22-044-653 is the replacement filter
Specifications
Product Type Replacement Filter
Filter Type Xylene Absorbing
For Use With (Application) Clean downdraft ductless fume hood