Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ SHUR/Clean™ Passive Open-Front Fume Hoods
SDP

Catalog No. 22047209 Shop All Fisherbrand Products
Change view
Click to view available options
Exhaust:
Rear Exhaust ports
Top Exhaust Ports
Width (Metric):
91 cm
122 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Exhaust Width (Metric)
22-047-209 Top Exhaust Ports 122 cm
22-047-207 Top Exhaust Ports 91 cm
22-047-208 Rear Exhaust ports 91 cm
22-047-210 Rear Exhaust ports 122 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-047-209

Without filters for installations to be externally exhausted

Exhaust ports are on the top or rear mounted. Can easily accomodate Microwave Tissue Processor and its accessories. (Contact your local architect and/or mechanical contractor to determine what accessories will be required to comply with local building codes.)

Requires ducting to HVAC system by user
Top (-TV) or rear (-RV) 6 in. O.D. exhaust ports

  • Accommodates the TBS SHUR/Wave microwave tissue processor
  • 36 in. Model
    • External dimensions: 36W x 26D x 28 in.H (91 x 66 x 71cm)
    • Internal dimensions: 34 x 25 x 27 in. (86 x 64 x 69cm)
    • Weight: 26 lb. (12kg)
    • Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047211): 4 large, 4 med. or 4-6 small bowls
    48 in. Model
    • External dimensions: 48W x 26D x 28 in.H (122 x 66 x 71cm)
    • Internal dimensions: 46 x 25 x 27 in. (117 x 64 x 69cm)
    • Weight: 32 lb. (15kg)
    • Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047212): 8 large, 8 med. or 10 small bowls

    Specifications

    Depth (English) 26 in.
    Depth (English) Interior 25 in.
    Depth (Metric) 66 cm
    Depth (Metric) Interior 64 cm
    Exhaust Top Exhaust Ports
    Height (English) 28 in.
    Height (English) Interior 27 in.
    Height (Metric) 71 cm
    Weight (English) 32 lb.
    Weight (Metric) 15 kg
    Width (English) 48 in.
    Width (English) Interior 46 in.
    Width (Metric) 122 cm
    Width (Metric) Interior 117 cm
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.