Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Small Round Biohazard Labels
GREENER_CHOICESDP

Catalog No. 18999931 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-999-931 Black, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-931
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

  • Strong adhesive ensures that labels remain in place
  • Red labels with black 1 in. core
  • Round, small, 1 in. dia.

Compliance

Meet OSHA requirements.

Specifications

Certifications/Compliance OSHA
Color Black, Red
Format Label
Material Paper
Product Type Round Biohazard Label
For Use With (Application) Biohazard
Quantity 1000/Roll
Shape Round
Diameter (English) 1.25 in.
Diameter (Metric) 3.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.