Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sparkleen™ 1 Detergent
SCI_ED

Catalog No. S701101 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3.25 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S701101 3.25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S701101

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sparkleen 1 is a universal detergent for manual or hand washing laboratory glassware and equipment. This mild highly buffered product is an excellent soak cleaner for most materials. Glassware washed with Sparkleen 1 is truly sparkling clean, entirely free from washing film, even with the hardest water.

 

  • Concentrated
  • Contains phosphates
  • Supplied as 3.25 lb.

 

Specifications

Form Powder
pH 9.5 to 10.2
Appearance White
Composition Sodium Carbonate 10 to 25%, Sodium Dodecylbenzenesulfonate 1 to 10%, non ionic detergent 1 to 10%
For Use With (Application) Removes Proteins, Fats, Oils, media, wax pencil marks, dried Blood and other organic-inorganic deposits from Glassware
Quantity 3.25 lb.
Volume (English) 3.25 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.