Learn More
Fisherbrand™ Speci-Freez™ Insulated Shipper
Catalog No. 22043208
Change view
Click to view available options
Type:
Insulated Shipper Kit
Refurbishing Kit
Replacement Outer Box
Quantity:
10/Cs.
4/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-043-208 Refurbishing Kit 4/Cs.
19-075-384C Insulated Shipper Kit 4/Cs.
22-043-207 Replacement Outer Box 10/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-043-208 Supplier Fisherbrand™ Supplier No. INF7050
Add to Cart
Add to Cart
Includes
Infectious Substance and Dry Ice Super Labels
Designed for use with the Infecon™ 2000, 2050 and 3000 shippers
  • For shipping infectious substances or diagnostic specimens that need to be shipped on dry ice
  • Durable, lightweight
  • Holds approximately 22 lb. (10kg) of dry ice, keeping contents frozen for up to 76 hours
Specifications
Quantity 4/Cs.
Applications For shipping infectious substances or diagnostic specimens that need to be shipped on dry ice
Type Refurbishing Kit
Dimensions (L x W x H) Exterior 15.5 x 14 x 12.75 in. (39.4 x 35.6 x 32.4 cm)
Dimensions (D x W x H) Interior 12.5 x 10.25 x 8 in.