Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Acid/Alkali Test Papers

Catalog No. 14861 Shop All Fisherbrand Products
Click to view available options
pH Range:
<3: Blue, >5.2: Red
0 to 14
0 to 7
7 to 14
Type:
Blue Litmus Test Strip
Congo Red Test Strip
Litmus Neutral Test Strip
Litmus Red Test Strip
This item is not returnable. View return policy
This item is not returnable. View return policy

Foolproof pH test to determine if a solution is acidic or alkaline.

  • A foolproof pH test to determine if a solution is acidic
  • The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
  • The test strip will turn red if dipped into an acidic solution (pH <7)
  • It will remain unchanged in an alkaline solution
  • We have tested the blue litmus with multiple pH buffers
  • The blue paper is blue at pH 8
  • At pH 7, the sample is maybe just a little red
  • The real change to red seems to start with pH 6
  • 5 and lower
  • Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
  • The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
  • Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
  • The test strip will turn blue if dipped into an alkaline solution (pH >7)
  • Color chart not included

Order Info

100 Strips per vial

Specifications

Certifications/Compliance OSHA, 29 CFR 1910.133, EN166
pH Range <3: Blue, >5.2: Red
Quantity 24 x 100 Strips
Type Congo Red Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.