Learn More
Fisherbrand™ Square Labels
GREENER_CHOICESDP
Catalog No. 11010244 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
Catalog No. Color
11-010-244 White
1 options
Catalog No. 11-010-244
Add to Cart
Add to Cart
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Specifications
Color White
Product Type Square label roll
Sterility Non-sterile
Quantity 2000/Roll
Size (English) 0.25 x 1.5 in.
Size (Metric) 0.64 x 3.81 cm