Learn More
Fisherbrand™ Square Labels
GREENER_CHOICESDP
Catalog No. 11010264 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
Catalog No. Color
11-010-264 White
1 options
Catalog No. 11-010-264
Add to Cart
Add to Cart
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Specifications
Color White
Product Type Square label roll
Sterility Non-sterile
Quantity 5000/Roll
Size (English) 0.75 x 1 in.
Size (Metric) 1.91 x 2.54 cm