Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ StainFrame Slide Holder
SDP

Catalog No. 2350050 Shop All Fisherbrand Products
Change view
Click to view available options
Slide Capacity:
14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
23-500-50 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-500-50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stain, drain and rinse up to 14 slides at once

  • Unique design allows user to rinse or decolorize slides with crystal-clear slide clarity and consistent results
  • Durable, chemical-resistant ABS plastic construction with easy-grip handle for complete control through the rinsing and decolorization process
  • Staining procedures can be performed with less mess and reagent waste than the Coplin jar method
  • Frame can be loaded with slides at the workstation for transport to the staining area
  • Slides are completely safe from breakage once the StainFrame is locked into position
  • Bottom frame may be used as a conventional staining rail

Specifications

Slide Capacity 14
Dimensions (L x W x H) 24 x 4 x 2 in. (61 x 10.2 x 5 cm)
For Use With (Equipment) Microscope Slide
Height (English) 2 in.
Height (Metric) 5 cm
Includes Easy-grip handle
Length (English) 24 in.
Length (Metric) 61 cm
Material ABS Plastic
Width (English) 4 in.
Width (Metric) 10.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.