Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Standard Grade Split Cowhide Leather Double Palm Gloves

Catalog No. 19816873 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Gray, Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-816-873 Gray, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-816-873
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from standard grade shoulder split cowhide leather, with double layered palms for additional comfort and protection.

  • Standard grade shoulder split cowhide leather construction
  • Double layered palm
  • Cotton lined for comfort
  • Gunn cut
  • Wing thumb

Specifications

Color Gray, Blue
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Universal
Style Gunn
Glove Cuff Style Safety
Liner Material Cotton
Thumb Style Wing
Length (English) 10.5 in.
Length (Metric) 26.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.