Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Standard Pipette Tips
GREENER_CHOICE

Catalog No. p-3206508 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-681-2 1 to 200 μL
02-681-4 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-681-2
Only null left
Add to Cart
Add to Cart

Standard tips for pipetters up to 1000 μL

For use with, but not limited to:

  • Costar™ Onepette™, Costar 8/12 Pette™
  • Eppendorf™
  • Thermo Scientific Finnpipette
  • LabPette™ 8- and 12-Channel
  • Nichiryo™
  • Pipetman™
  • Rainin™ EDP2™
  • Oxford™ Benchmate™
  • Socorex™
  • Titertek Digital™
  • UNITEK™ Volac™
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Color Yellow
Tip Style Universal
Packaging Bulk Packed
For Use With (Equipment) Gilson Pipetman, Costar Onepette, Costar 8-pette, Costar 12-pette, Eppendorf, Thermo Scientific Finnpipette, Labpette Multichannel, Nichiryo, Oxford Benchmate, Rainin EDP2, Titertek
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.