Learn More
Fisherbrand™ StayRite™ Pointer for StayRite Hydrometers
SDP
Catalog No. 13510PNTR Shop All Fisherbrand Products
Catalog No. 13-510-PNTR
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
StayRite Pointer. Used to rotate hydrometer's internal magnet to its proper position. Can be used to set any StayRite instrument.