Learn More
Fisherbrand™ Super Friendly Freeze'It™
Catalog No. 23022524 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Freeze Spray
Catalog No. Product Type
23-022524 Freeze Spray
1 options
Catalog No. 23-022524
Includes
11 oz. (312g) of noncorrosive, nonflammable, odorless, completely volatile gas; 5 in. (12.7cm) polyethylene snorkel
Aerosol for cooling liquid, semisolid, and solid samples.
Aerosol for cooling liquid, semisolid, and solid samples
  • Rapidly cools any material without CFCs or HCFCs
  • Concentrates freezing power exactly where needed
  • Capable of dropping sample temperature from room temperature to -51°C (-60°F) in three seconds
  • Triple-filtered gas prevents sample contamination and enhances cooling ability
  • Deters the water crystal formation that occurs with slower refrigerants
Specifications
Applications Cooling liquid, Semisolid and Solid samples
Volume (English) 2 oz.
Product Type Freeze Spray