Learn More
Fisherbrand™ Superfrost™ Excell™ Microscope Slides
Catalog No. 22034985 Shop All Fisherbrand Products
Change view
Click to view available options
Corner Style:
Regular
Catalog No. Corner Style
22-034-985 Regular
1 options
This item is not returnable. View return policy
Catalog No. 22-034-985
This item is not returnable. View return policy
New, innovative adhesive technology for superior adhesion and wettability
  • Developed specifically for immunohistochemistry laboratory applications
  • Adhesion surface for heat-induced epitope retrieval
  • For use in HIER methods requiring high pH antigen retrieval solutions, including EDTA
  • Uniform hydrophilic surface works well with automated staining equipment
  • Constructed of glass to work well with both epon and methyl methacrylate sections
  • Hydrophilic surface allows sections to spread well during initial attachment; section adherence is maintained throughout difficult staining procedures
  • Excellent tissue adherence of all tissue types, yet easily release identified areas for Laser Capture Microdissection
  • Enhanced attachment of anchorage dependent cell lines, including many fastidious lines, e.g., primary neuronal cells
Specifications
Material Glass
Frosted Yes
Type Microscope Slide
Coating Adhesion
Corner Style Regular