Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SURE-TEK™ 2 Biopsy Cassette Taped Sleeves
SDP

Catalog No. p-4670114 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-048-111 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-048-111
This item is not returnable. View return policy

Used in Leica Cassette Labelers

  • Comes with attached lids
  • Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac color

Specifications

For Use With (Application) Leica Cassette Labelers
Includes Attached lid
Type Biopsy Cassette Taped Sleeve
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.