Learn More
Fisher Healthcare™ SURE-TEK™ 2 Processing and Embedding Cassette Taped Sleeves
Shop All Fisherbrand Products

Description
- Comes with attached lids
- Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac color
Specifications
Specifications
| For Use With (Application) | Leica Cassette Labelers |
| Includes | For table or wall mounting |
| Type | Processing and Embedding Cassette Taped Sleeve |
| Color | Blue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.