Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SURE-TEK™ 2 Processing and Embedding Cassette Taped Sleeves
SDP

Catalog No. 22048105 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Pink
Includes:
Attached lid
For table or wall mounting
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
22-048-105 Pink Attached lid
22-048-102 Blue For table or wall mounting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-048-105

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in Leica Cassette Labelers

  • Comes with attached lids
  • Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac color

Specifications

For Use With (Application) Leica Cassette Labelers
Includes Attached lid
Type Processing and Embedding Cassette Taped Sleeve
Color Pink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.