Learn More
Fisher Healthcare™ SURE-TEK™ 2 Processing Embedding Cassettes Dispenser Packs
Shop All Fisherbrand Products

Description
- Supplied in Dispenser Packs
- Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac colors
Specifications
Specifications
| For Use With (Equipment) | Sure-tek™ 2 Cassette |
| Includes | Attached lid |
| Type | Processing and Embedding Cassette |
| Color | Peach |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.