Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Surface Protectors in Rolls

Catalog No. 14666300 Shop All Fisherbrand Products
Change view
Click to view available options
Width (English):
18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
14-666-300 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-666-300

missing translation for 'validMainframeMsg'

Polyshield fluid barrier is a super absorbent for splatters and spills, and protects work areas from contamination and damage by toxic, infectious, corrosive, radioactive or staining materials

  • Pure cellulose paper
  • Impermeable polyethylene backing
  • Smooth, firm surface
  • Does not inhibit stability of bottles or tubes, and allows recovery of dry spills
  • Disposal is safe and easy
  • Polyshield ashes readily upon incineration

Liquid samples can be recovered by washing the absorbed spill from the inert surface with the appropriate buffer.

Specifications

Product Type Bench Protector
Material Cellulose/Polyethylene
Length (English) 164 ft.
Length (Metric) 50 m
Width (English) 18 in.
Width (Metric) 46 m
For Use With (Application) Polyshield fluid barrier is a super absorbent for splatters and spills, and protects work areas from contamination and damage by toxic, infectious, corrosive, radioactive or staining materials
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.