Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
SCI_ED

Catalog No. p-4273780 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
1 Stainless Steel Probe
2 Stainless Steel Probes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S04149 2 Stainless Steel Probes
S04148 1 Stainless Steel Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S04149
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Probe(s), two AA batteries, Stand, Velcro™ magnetic strips, wall mount, Traceable™ certificate

Monitors inlet and outlet simultaneously. Traceable™ Hi-Accuracy Thermometer records time and date for MIN/MAX readings.

  • Two visual (LEDs) and two audio alarm alerts
  • Alarms programmable in 0.1° increments
  • Probe tip penetration of 0.33 in.
  • Stainless-steel probe with 0.125 in. diameter, 6.25 in. stem length, 9 in. overall length and 5 ft. cable length
  • Available with one probe or two probes
  • Includes 2 AA Batteries, Stand, Velcro™ Magnetic Strips, Wall Mount, Traceable™ Certificate

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Compliance

Conforms to ITS-90, International Temperature Standard

Specifications

Certifications/Compliance NIST, ITS-90, ISO 17025, A2LA
Depth (English) 0.75 in.
Depth (Metric) 1.9 cm
Diameter (English) Probe 0.12 in.
Diameter (Metric) Probe 3.05 mm
Dimensions (L x W x H) 9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm)
For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater
Height (English) 4.25 in.
Height (Metric) 10.79 cm
Includes 2 Stainless Steel Probes
Length (English) Cable 5 ft.
Length (English) Probe 6.25 in.
Length (Metric) 230 mm
Length (Metric) Cable 152.4 cm
Length (Metric) Probe 15.87 cm
Material Plastic
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 141.7 g
Width (English) 2.75 in.
Product Type Traceable Thermometer
Length (English) 9 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58° to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Battery Type AA
Accuracy ±0.3°C
Display LED
Width (Metric) 6.98 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.