Learn More
Fisherbrand™ Ultrasonic Units Cooling Coils
SDP
Catalog No. 1038735 Shop All Fisherbrand Products
Change view
Click to view available options
Model:
S30 - S70
S80 - S300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
1038735 S30 - S70
1037515 S80 - S300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1038735
Add to Cart
Add to Cart
The Fisherbrand™ Ultrasonic Unit Cooling Coil is designed specifically for use with the Fisherbrand™ 112xx Series of dual frequency ultrasonic cleaners. Maintaining a stabile liquid temperature is critical in ultrasonic cleaning.
  • Keep the cooling bath temperature stable
  • Ensure proper ultrasonic cleaning with the exact temperature
  • Connects to the closed cycle cooling system
Specifications
Type Cooling Coil
Diameter (Metric) Inner 6 mm
Diameter (Metric) 8 mm
For Use With (Equipment) Elmasonic Unit Sizes: S 30, S 40, S 70, S 150
Material Stainless Steel
Model S30 - S70