Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
SDP

Catalog No. FB15083 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, 18 L
Ultrasonic Cleaner, 2.75 and 4.25 L
Ultrasonic Cleaner, 28 L
Ultrasonic Cleaner, 5.75 L
Ultrasonic Cleaner, 6.9 and 9.4 L
Ultrasonic Cleaner, 9.5 and 12.75 L
Type:
Lid
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
FB15083 Ultrasonic Cleaner, 2.75 and 4.25 L Lid
FB15084 Ultrasonic Cleaner, 5.75 L Lid
FB15086 Ultrasonic Cleaner, 9.5 and 12.75 L Lid
FB15087 Ultrasonic Cleaner, 18 L Lid
FB15088 Ultrasonic Cleaner, 28 L Lid
FB15085 Ultrasonic Cleaner, 6.9 and 9.4 L Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. FB15083
Only null left
Add to Cart
Edge
Add to Cart

A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.

  • Easy-to-open and close cover
  • Muffles noise level
  • Designed to be used as drip basin when flipped over
  • Chemical-resistant plastic
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage

Specifications

Type Lid
For Use With (Equipment) Ultrasonic Cleaner, 2.75 and 4.25 L
For Use With (Application) Medical Laboratories and in Industrial Production Processes
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.